Hydrogen peroxide 30% stabilized, BAKER ANALYZED® ACS, J.T.Baker®

Brand: Avantor Performance Materials, LLC
Danger


Management of Change (MOC) category = N

These acids are tested for common trace metals.

Formula: H₂O₂
MW: 34.01 g/mol
Boiling Pt: 107 °C (1013 hPa)
Melting Pt: ? 0 °C
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1 (8),II

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (H₂O₂) 30.0 - 32.0 %
Color (APHA) ≤10
Residue after Evaporation ≤0.0020 %
Titrable Acid (meq/g) ≤0.0006
Chloride (Cl) ≤2 ppm
Nitrate (NO₃) ≤2 ppm
Phosphate (PO₄) ≤2 ppm
ACS - Sulfate (SO₄) ≤5 ppm
Ammonium (NH₄) ≤5 ppm
Trace Impurities - Copper (Cu) ≤50 ppb
Trace Impurities - Iron (Fe) ≤100.0 ppb
Heavy Metals (as Pb) ≤1000.0 ppb
Trace Impurities - Lead (Pb) ≤100 ppb
Trace Impurities - Nickel (Ni) ≤50 ppb
Meets Reagent Specifications for testing USP/NF monographs