MSDS Number: B5641 * * * * * Effective Date: 10/01/08 * * * * * Supercedes: 04/21/08 Buffer Solution (Biphthalate), pH 4 (Color Coded Red)
MSDS   MATERIAL SAFETY DATA SHEET                  CHEMTREC: 800-424-9300 (USA)
              ---------------------------                    703-527-3887(Outside USA and Canada)
                                                                                              CANUTEC:    613-996-6666
From:    Mallinckrodt Baker, Inc
              222 Red School Lane
              Phillipsburg, NJ 08865                                  NOTE: Use CHEMTREC and CANUTEC
                                                                                                  phone numbers only in the event
Emergency Telephone Number: 908-859-2151                        of a chemical emergency.
All non-emergency questions should be directed to Customer Service (1-800-582-2537) for assistance.
M A L L I N C K R O D T                                      J. T. B A K E R
Buffer Solution (Biphthalate), pH 4 (Color Coded Red)
1. Product Identification
Synonyms: None.
CAS No.: Not applicable to mixtures.
Molecular Weight: Not applicable to mixtures.
Chemical Formula: Not applicable to mixtures.
Product Codes:
J.T. Baker: 5657
Mallinckrodt: 0097
2. Composition/Information on Ingredients
  Ingredient                                CAS No         Percent        Hazardous
  ---------------------------------------   ------------   ------------   ---------
  Water                                     7732-18-5        98 - 99%        No
  Potassium Acid Phthalate                  877-24-7          1 - 2%         Yes
  Thymol                                    89-83-8            < 1%          Yes
  FD & C Red No. 40                         25956-17-6         < 1%          No
3. Hazards Identification
Emergency Overview
--------------------------
CAUTION! MAY CAUSE IRRITATION TO SKIN AND EYES.
SAF-T-DATA(tm) Ratings (Provided here for your convenience)
-----------------------------------------------------------------------------------------------------------
Health Rating: 1 - Slight
Flammability Rating: 0 - None
Reactivity Rating: 0 - None
Contact Rating: 2 - Moderate
Lab Protective Equip: GOGGLES; LAB COAT; PROPER GLOVES
Storage Color Code: Green (General Storage)
-----------------------------------------------------------------------------------------------------------
Potential Health Effects
----------------------------------
Information on the human health effects from exposure to this substance is limited.
Inhalation:
Not expected to be an inhalation hazard. May cause irritation to respiratory tract because of slight acidity. Symptoms may include coughing and sore throat.
Ingestion:
Large doses may produce nausea, vomiting, and abnormal sensations in hands and feet. Because of slight acidity, causes irritation to the mucous membranes.
Skin Contact:
Contact may cause irritation, with redness and pain.
Eye Contact:
May cause eye irritation.
Chronic Exposure:
No information found.
Aggravation of Pre-existing Conditions:
No information found.
4. First Aid Measures
Inhalation:
Remove to fresh air. Get medical attention for any breathing difficulty.
Ingestion:
If large amounts were swallowed, give water to drink and get medical advice.
Skin Contact:
Immediately flush skin with plenty of water for at least 15 minutes. Remove contaminated clothing and shoes. Wash clothing before reuse. Thoroughly clean shoes before reuse. Get medical attention if irritation develops.
Eye Contact:
Immediately flush eyes with plenty of water for at least 15 minutes, lifting upper and lower eyelids occasionally. Get medical attention if irritation persists.
5. Fire Fighting Measures
Fire:
Not expected to be a fire hazard.
Explosion:
No information found.
Fire Extinguishing Media:
Use any means suitable for extinguishing surrounding fire.
Special Information:
In the event of a fire, wear full protective clothing and NIOSH-approved self-contained breathing apparatus with full facepiece operated in the pressure demand or other positive pressure mode.
6. Accidental Release Measures
Ventilate area of leak or spill. Wear appropriate personal protective equipment as specified in Section 8. Contain and recover liquid when possible. Collect liquid in an appropriate container or absorb with an inert material (e. g., vermiculite, dry sand, earth), and place in a chemical waste container. Do not use combustible materials, such as saw dust.
7. Handling and Storage
Keep in a tightly closed container, stored in a cool, dry, ventilated area. Protect against physical damage. Containers of this material may be hazardous when empty since they retain product residues (vapors, liquid); observe all warnings and precautions listed for the product.
8. Exposure Controls/Personal Protection
Airborne Exposure Limits:
None established.
Ventilation System:
In general, dilution ventilation is a satisfactory health hazard control for this substance. However, if conditions of use create discomfort to the worker, a local exhaust system should be considered.
Personal Respirators (NIOSH Approved):
Not expected to require personal respirator usage.
Skin Protection:
Wear protective gloves and clean body-covering clothing.
Eye Protection:
Use chemical safety goggles. Maintain eye wash fountain and quick-drench facilities in work area.
9. Physical and Chemical Properties
Appearance:
Clear, reddish liquid.
Odor:
Odorless.
Solubility:
Completely soluble in water.
Specific Gravity:
No information found.
pH:
4.0
% Volatiles by volume @ 21C (70F):
ca. 98
Boiling Point:
No information found.
Melting Point:
No information found.
Vapor Density (Air=1):
No information found.
Vapor Pressure (mm Hg):
No information found.
Evaporation Rate (BuAc=1):
No information found.
10. Stability and Reactivity
Stability:
Stable under ordinary conditions of use and storage.
Hazardous Decomposition Products:
Carbon dioxide and carbon monoxide may form when heated to decomposition.
Hazardous Polymerization:
Will not occur.
Incompatibilities:
No information found.
Conditions to Avoid:
Heat, incompatibles.
11. Toxicological Information
No LD50/LC50 information found relating to normal routes of occupational exposure.
  --------\Cancer Lists\------------------------------------------------------
                                         ---NTP Carcinogen---
  Ingredient                             Known    Anticipated    IARC Category
  ------------------------------------   -----    -----------    -------------
  Water (7732-18-5)                       No          No            None
  Potassium Acid Phthalate (877-24-7)     No          No            None
  Thymol (89-83-8)                        No          No            None
  FD & C Red No. 40 (25956-17-6)          No          No            None
12. Ecological Information
Environmental Fate:
No information found.
Environmental Toxicity:
No information found.
13. Disposal Considerations
Whatever cannot be saved for recovery or recycling should be managed in an appropriate and approved waste disposal facility. Processing, use or contamination of this product may change the waste management options. State and local disposal regulations may differ from federal disposal regulations. Dispose of container and unused contents in accordance with federal, state and local requirements.
14. Transport Information
Not regulated.
15. Regulatory Information
  --------\Chemical Inventory Status - Part 1\---------------------------------
  Ingredient                                       TSCA  EC   Japan  Australia
  -----------------------------------------------  ----  ---  -----  ---------
  Water (7732-18-5)                                 Yes  Yes   Yes      Yes
  Potassium Acid Phthalate (877-24-7)               Yes  Yes   Yes      Yes
  Thymol (89-83-8)                                  Yes  Yes   Yes      Yes
  FD & C Red No. 40 (25956-17-6)                    Yes  Yes   No       Yes
  --------\Chemical Inventory Status - Part 2\---------------------------------
                                                          --Canada--
  Ingredient                                       Korea  DSL   NDSL  Phil.
  -----------------------------------------------  -----  ---   ----  -----
  Water (7732-18-5)                                 Yes   Yes   No     Yes
  Potassium Acid Phthalate (877-24-7)               Yes   Yes   No     Yes
  Thymol (89-83-8)                                  Yes   Yes   No     Yes
  FD & C Red No. 40 (25956-17-6)                    Yes   Yes   No     Yes
  --------\Federal, State & International Regulations - Part 1\----------------
                                             -SARA 302-    ------SARA 313------
  Ingredient                                 RQ    TPQ     List  Chemical Catg.
  -----------------------------------------  ---   -----   ----  --------------
  Water (7732-18-5)                          No    No      No         No
  Potassium Acid Phthalate (877-24-7)        No    No      No         No
  Thymol (89-83-8)                           No    No      No         No
  FD & C Red No. 40 (25956-17-6)             No    No      No         No
  --------\Federal, State & International Regulations - Part 2\----------------
                                                        -RCRA-    -TSCA-
  Ingredient                                 CERCLA     261.33     8(d)
  -----------------------------------------  ------     ------    ------
  Water (7732-18-5)                          No         No         No
  Potassium Acid Phthalate (877-24-7)        No         No         No
  Thymol (89-83-8)                           No         No         No
  FD & C Red No. 40 (25956-17-6)             No         No         No
Chemical Weapons Convention:  No     TSCA 12(b):  No     CDTA:  No
SARA 311/312:  Acute: Yes      Chronic: No   Fire: No  Pressure: No
Reactivity: No          (Mixture / Liquid)
Australian Hazchem Code: None allocated.
Poison Schedule: None allocated.
WHMIS:
This MSDS has been prepared according to the hazard criteria of the Controlled Products Regulations (CPR) and the MSDS contains all of the information required by the CPR.
16. Other Information
NFPA Ratings: Health: 1 Flammability: 0 Reactivity: 0
Label Hazard Warning:
CAUTION! MAY CAUSE IRRITATION TO SKIN AND EYES.
Label Precautions:
Avoid contact with eyes, skin and clothing.
Keep container closed.
Wash thoroughly after handling.
Label First Aid:
In case of contact, immediately flush eyes or skin with plenty of water for at least 15 minutes. Get medical attention if irritation develops or persists.
Product Use:
Laboratory Reagent.
Revision Information:
MSDS Section(s) changed since last revision of document include: 1, 2, 9, 15.
Disclaimer:
************************************************************************************************
Mallinckrodt Baker, Inc. provides the information contained herein in good faith but makes no representation as to its comprehensiveness or accuracy. This document is intended only as a guide to the appropriate precautionary handling of the material by a properly trained person using this product. Individuals receiving the information must exercise their independent judgment in determining its appropriateness for a particular purpose. MALLINCKRODT BAKER, INC. MAKES NO REPRESENTATIONS OR WARRANTIES, EITHER EXPRESS OR IMPLIED, INCLUDING WITHOUT LIMITATION ANY WARRANTIES OF MERCHANTABILITY, FITNESS FOR A PARTICULAR PURPOSE WITH RESPECT TO THE INFORMATION SET FORTH HEREIN OR THE PRODUCT TO WHICH THE INFORMATION REFERS. ACCORDINGLY, MALLINCKRODT BAKER, INC. WILL NOT BE RESPONSIBLE FOR DAMAGES RESULTING FROM USE OF OR RELIANCE UPON THIS INFORMATION.
************************************************************************************************
Prepared by: Environmental Health & Safety
Phone Number: (314) 654-1600 (U.S.A.)